Kimble™ KIMAX™ Class A To Contain Graduated Cylinders
Actual product may vary
DWK Life Sciences
Kimble™ KIMAX™ Class A To Contain Graduated Cylinders
With bumpers and reverse metric scales
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS08-548-204100 mL1 mL
FIS08-548-20110 mL0.1 mL
FIS08-548-2071000 mL10 mL
FIS08-548-2082000 mL20 mL
Catalog number FIS08-548-204
Price (SGD)
915.00
Case
Capacity (Metric):
100 mL
Graduations:
1 mL
Price (SGD)
915.00
Case
  • Calibrated to contain at 20°C
  • Provided with SAFE-GUARD™ bumpers (25mL and larger)
  • Hexagonal bases are flat ground for added stability
  • Designed from ASTM E1272 Style 1, Class A requirements
  • Frequently used as receivers for distillation procedures
Specifications
MaterialBorosilicate Glass
Capacity (Metric)100 mL
IncludesSafe Gard Bumper
ScaleReverse metric scale
Graduations1 mL
Subdivision5 to 100 mL
CalibrationTo Contain
Base TypeHexagonal
For Use With (Equipment)Safe Gard Bumper
ShapeRound
ClassClass A
Height (English)10.03 in.
Height (Metric)255 mm
Certifications/ComplianceASTM E438, USP Type I
Tolerance±0.35 mL
Product TypeGraduated Measuring Cylinder
Unit SizeCase