PYREX™ Certified/Serialized Class A Mixing Cylinders, TC
PYREX™ Certified/Serialized Class A Mixing Cylinders, TC
Actual product may vary
PYREX™ Certified/Serialized Class A Mixing Cylinders, TC
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FIS08-531E250 mL2 mL
FIS08-531C50 mL1 mL
FIS08-531D100 mL1 mL
Catalog number FIS08-531E
Price (SGD)
676.00
CS
Add to cart
Capacity (Metric):
250 mL
Graduations:
2 mL
Price (SGD)
676.00
CS
Add to cart
Calibrated “To Contain” in accordance with ASTM E-1272 and ASTM E-542. Useful as a mixing cylinder. Each cylinder is individually serialized and supplied with a Certificate of Identification and Capacity, traceable to NIST standards. Each features the PYREX stopper. Graduation markings are in durable white enamel.
For stopper only, see Cat. No. 7650.
Tested and calibrated in an ISO/IEC 17025 certified laboratory. 
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorWhite
ShapeRound
SterilityNon-sterile
TypeGraduated Mixing Cylinder
Tolerance±1 mL
MaterialBorosilicate Glass
Certifications/ComplianceASTM E1272, ASTM E542
ClassA
Diameter (English) Outside1.53 in.
DisposableReusable
Capacity (Metric)250 mL
Diameter (Metric) Outside39 mm
For Use With (Application)Useful as a Mixing Cylinder
Height (English)15.15 in.
Height (Metric)385 mm
IncludesStopper
ScaleSingle
Stopper Number22
Stopper MaterialGlass
Stopper StyleHollow
Tolerance (Metric)±1 mL
Closure MaterialGlass
Graduations2 mL
AutoclavableAutoclavable
Subdivision2 mL
Unit SizeCS