Porcelain Combustion Boats
Actual product may vary
CoorsTek
Porcelain Combustion Boats
AD-998 High-Alumina labware
Have Questions?
Catalog NumberCapacity (Metric)Height (English)Length (English)Width (English)
FIS08-226-13F20 mL0.57 in.4.13 in.0.86 in.
Catalog number FIS08-226-13F
Price (SGD)
355.00
Each
Capacity (Metric):
20 mL
Height (English):
0.57 in.
Length (English):
4.13 in.
Width (English):
0.86 in.
Price (SGD)
355.00
Each
Specifications
Bottom ShapeFlat
Capacity (Metric)20 mL
MaterialAluminum, Porcelain
ShapeBoat
TypeCombustion Boat
For Use With (Application)For refractory metals such as molybdenum, platinum, rhodium, tungsten, tantalum and iridium
Dimensions (L x W x H)4.13 x 0.86 x 0.57 in. (105 x 22 x 14.5 mm)
Length (English)4.13 in.
Length (Metric)105 mm
Width (English)0.86 in.
Width (Metric)22 mm
Height (Metric)14.5 mm
Height (English)0.57 in.
Unit SizeEach