▸
Specialty Lab Equipment, Instruments, and Apparatuses
▸
Crucibles
▸
Fused Quartz Crucible Covers
Actual product may vary
QSI
Fused Quartz Crucible Covers
For 08-072 Series Crucibles
Have Questions?
Catalog Number
For Use With (Application)
Length (Metric)
FIS08-074A
For 10mL crucible
0.6cm
Catalog number
FIS08-074A
Price (SGD)
-
For Use With (Application):
For 10mL crucible
Length (Metric):
0.6cm
Transparent fused quartz
Specifications
Color
Clear, Transparent
Material
Quartz
For Use With (Application)
For 10mL crucible
Length (Metric)
0.6cm
Product Type
Crucible Lid