Mini LED Light Box
Mini LED Light Box
Actual product may vary
Mini LED Light Box
Mini LED Light Box
Fisherbrand
Mini LED Light Box
Backlight samples for easy viewing
Have Questions?
Catalog NumberDisplay Type
FIS07-910-023LED
Catalog number FIS07-910-023
Price (SGD)
568.00
EA
Add to cart
Display Type:
LED
Price (SGD)
568.00
EA
Add to cart
  • Enhances viewing of transparent and semi-transparent objects (96-well plates, microscope slides, petri dishes, electrophoresis gels and transparencies)
  • Cool-cathode LED source keeps viewing area cool to prevent sample damage
    • 10,000 hr. lifespan
  • Viewing area: 12.5 x 10 cm
  • Counting grid for segmented counting of sample regions
  • Adhesive strips provide semi-permanent grid attachment to light box
  • Vinyl storage case included
Notes:
  • Does not include required 6 AAA batteries or 9 V AC adapter
Specifications
Product TypeMini LED light box
Display TypeLED
Height (English)0.5 in.
Height (Metric)1.4 cm
Length (English)6.25 in.
Length (Metric)15.8 cm
Width (English)5.875 in.
Width (Metric)15 cm
For Use With (Application)Enhances viewing of transparent and semi-transparent objects
IncludesCounting grid and vinyl storage case
Unit SizeEA