Cork Stoppers - Laboratory Grade
Actual product may vary
Cork Stoppers - Laboratory Grade
100% natural cork
Have Questions?
Change viewbuttonViewtableView
Catalog NumberStopper NumberQuantity
FIS07-781P11100/Pk.
FIS07-781G4100/Pk.
FIS07-781L8100/Pk.
FIS07-781N10100/Pk.
Catalog number FIS07-781P
Price (SGD)
46.00
Pack
Stopper Number:
11
Quantity:
100/Pk.
Price (SGD)
46.00
Pack
100% natural cork
  • Punched out from natural cork wood
  • Tapered to size
  • Safe to use in the packaging and/or processing of food
Specifications
For Use With (Application)Ideal for sealing bottles, safe to use in the packaging and/or processing of food
GradeLaboratory
Length (Metric)3.17 cm
Quantity100/Pk.
Size11
ColorNatural
Diameter (Metric) Bottom2.1 cm
Diameter (Metric) Top1.75 cm
MaterialWood
Product TypeCork Stopper
Stopper Number11
Unit SizePack