Drying tube for air and gases. W A Hammond Drierite™ Drying Tubes dries air and gases for small scale laboratory applications and contains a flexible hose connection provides an immediate flow of dry gas.
Have Questions?
Catalog Number
Quantity
FIS07-578-5
4 Tubes
Catalog number FIS07-578-5
Price (SGD)
201.00
EA
Add to cart
Quantity:
4 Tubes
Price (SGD)
201.00
EA
Add to cart
Drying tube for air and gases. W A Hammond Drierite™Drying Tubes dries air and gases for small scale laboratory applications and contains a flexible hose connection provides an immediate flow of dry gas.
Vent drier on reagent bottles, reaction apparatus, or instrument compartment,
For sample preparation, gravimetric determination of water vapor, drying of gases for purging and blanketing of moisture sensitive materials
Contains flexible hose connection, provides immediate flow of dry gas
Turns from blue to pink to give constant visual assurance of active desiccant
Polycarbonate end caps
20μm polypropylene filter in each end
Dimensions (O.D x Length): 1.9 x 15.2cm (0.75x 6 in.)
Specifications
Chemical Name or MaterialDrierite Indicating Drying Tube, WA Hammond