Drierite™ Indicating Absorbents, W.A. Hammond
Drierite™ Indicating Absorbents, W.A. Hammond
Drierite™ Indicating Absorbents, W.A. Hammond
Drierite™ Indicating Absorbents, W.A. Hammond
W.A. Hammond
Drierite™ Indicating Absorbents, W.A. Hammond
Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 lb.
1 lb.
Mesh Size:
8
Catalog number FIS07-578-3B
Price (SGD)
422.00
EA
Add to cart
Quantity:
5 lb.
Mesh Size:
8
Request bulk or custom format
Price (SGD)
422.00
EA
Add to cart
Chemical Identifiers
CAS7646-79-9 , 7778-18-9
Specifications
Chemical Name or MaterialCalcium Sulfate with Cobalt Chloride Indicator
CAS7646-79-9
CAS7778-18-9
MaterialCalcium Sulfate with Cobalt Chloride
Quantity5 lb.
View more
Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion. W A Hammond Drierite™ Drying Dessicants dries organic liquids to low ppm levels and is commonly used in desiccators, desiccator cabinets, in-line air and gas dryers.
  • Efficient desiccant giving constant visual indication of the progress of absorption and exhaustion
  • Turns from blue to pink when exhausted
  • Dries air and gases to lower residual moisture levels (-100°F dew point)
  • Commonly used in desiccators, desiccator cabinets, in-line air and gas drying columns, absorption bulbs, U-tubes, glove boxes , sealed chambers and wherever an anhydrous environment is required
  • Regeneration by heating granules on tray in single layer at 210-220°C for 1 hour