Search Fisher Scientific
| Catalog Number | Capacity (Metric) |
|---|---|
| FIS07-250-071 | 500 mL |
| FIS07-250-066 | 10 mL |
| FIS07-250-067 | 25 mL |
| FIS07-250-068 | 50 mL |
| FIS07-250-069 | 100 mL |
| FIS07-250-070 | 250 mL |
Calibrated To Deliver
These Class A graduated cylinders are calibrated 'To Deliver' and have blue enamel graduations. The cylinder capacity is in accordance with ASTM E-1272. The 10 mL size has a funnel top for easier filling.