PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TD
Actual product may vary
PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TD
Calibrated To Deliver
Have Questions?
Catalog NumberCapacity (Metric)
FIS07-250-070250 mL
Catalog number FIS07-250-070
Price (SGD)
129.00
Each
Capacity (Metric):
250 mL
Price (SGD)
129.00
Each
Calibrated To Deliver
These Class A graduated cylinders are calibrated 'To Deliver' and have blue enamel graduations. The cylinder capacity is in accordance with ASTM E-1272. The 10 mL size has a funnel top for easier filling.
Specifications
MaterialBorosilicate Glass
Capacity (Metric)250 mL
ScaleSingle
AutoclavableAutoclavable
SterilityNon-sterile
CalibrationTo Deliver
Base TypeHexagonal
Scale ColorBlue
ShapeRound
ClassClass A
Height (English)13.38 in.
Height (Metric)340 mm
Certifications/ComplianceASTM E1272, ASTM E542, ASTM E694
Tolerance±1 mL
Product TypeGraduated Cylinder
Quantity1/Pk.
Diameter (Metric) Outer39 mm
Unit SizeEach