PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TC
Actual product may vary
PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TC
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS07-250-063100 mL
FIS07-250-06250 mL
Catalog number FIS07-250-063
Price (SGD)
94.00
Pack
Capacity (Metric):
100 mL
Price (SGD)
94.00
Pack
These Class A graduated cylinders are calibrated 'to contain' and have blue enamel graduations. The cylinder capacity is in accordance with ASTM E-1272. The 10 mL size has a funnel top. For bumper guard only, see Cat. No. 3066.
Specifications
Base TypeHexagonal
CalibrationTo Contain
Scale ColorBlue
ShapeRound
SterilityNon-sterile
Certifications/ComplianceASTM E1272, ASTM E542, ASTM E694
ClassA
Height (Metric)254 mm
Tolerance Metric±0.5 mL
TypeGraduated Cylinder
MaterialBorosilicate Glass
Diameter (Metric) Outside29 mm
Capacity (Metric)100 mL
IncludesFunnel Top, Pourout
ScaleSingle