PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TC
Actual product may vary
PYREX™ VISTA™ Class A Graduated Cylinder, Single Metric Scale, TC
Calibrated To Contain
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FIS07-250-06250 mL
FIS07-250-063100 mL
Catalog number FIS07-250-062
Price (SGD)
79.00
Pack
Capacity (Metric):
50 mL
Price (SGD)
79.00
Pack
These Class A graduated cylinders are calibrated 'to contain' and have blue enamel graduations. The cylinder capacity is in accordance with ASTM E-1272. The 10 mL size has a funnel top. For bumper guard only, see Cat. No. 3066.
Specifications
MaterialBorosilicate Glass
Capacity (Metric)50 mL
IncludesFunnel Top, Pourout
ScaleSingle
AutoclavableAutoclavable
SterilityNon-sterile
CalibrationTo Contain
Base TypeHexagonal
Scale ColorBlue
ShapeRound
ClassClass A
Height (Metric)225 mm
Certifications/ComplianceASTM E1272, ASTM E542, ASTM E694
Tolerance±0.25 mL
Product TypeGraduated Cylinder
Diameter (Metric) Outer24 mm
Unit SizePack