Polypropylene Screw Cap Dipper With Removable Handle, Blue, Sterile
Polypropylene Screw Cap Dipper With Removable Handle, Blue, Sterile
Actual product may vary
Polypropylene Screw Cap Dipper With Removable Handle, Blue, Sterile
Quality sampling tools for secure sample retrieval and shipment
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (English)Height (Metric)
FIS07-202-03240 mL10.8 in.275 mm
FIS07-201-97190 mL11.41 in.29 cm
Catalog number FIS07-202-032
Price (SGD)
1,335.00
Case
Add to cart
Capacity (Metric):
40 mL
Height (English):
10.8 in.
Height (Metric):
275 mm
Price (SGD)
1,335.00
Case
Add to cart
  • Polypropylene blue containers
  • Removable handle
  • Shock resistant, Leakproof
  • No cross contamination risks
  • Traceability information on each bag
  • Packaged individually in tamper evident single zipped bags
Specifications
Product TypeScrew Cap Dipper
MaterialPolypropylene
Handle TypeRemovable
Capacity (Metric)40 mL
SterilitySterile
ColorBlue
Height (English)10.8 in.
Height (Metric)275 mm
Unit SizeCase