Polypropylene Screw Cap Dipper With Removable Handle, Blue, Sterile
Actual product may vary
Polypropylene Screw Cap Dipper With Removable Handle, Blue, Sterile
Quality sampling tools for secure sample retrieval and shipment
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Height (English)Height (Metric)
FIS07-201-97190 mL11.41 in.29 cm
FIS07-202-03240 mL10.8 in.275 mm
Catalog number FIS07-201-971
Price (SGD)
968.00
Case
Capacity (Metric):
90 mL
Height (English):
11.41 in.
Height (Metric):
29 cm
Price (SGD)
968.00
Case
  • Polypropylene blue containers
  • Removable handle
  • Shock resistant, Leakproof
  • No cross contamination risks
  • Traceability information on each bag
  • Packaged individually in tamper evident single zipped bags
Specifications
Product TypeDipper
MaterialPolypropylene
IncludesRemovable handle
Capacity (English)3.04 oz.
Capacity (Metric)90 mL
SterilitySterile
ColorBlue
Certifications/ComplianceEC N 1935/2004, ISO 11137
For Use With (Application)Quality sampling tools for secure sample retrieval and shipment
Diameter (English) Outer1.69 in.
Diameter (Metric) Outer4.3 cm
Height (English)11.41 in.
Height (Metric)29 cm
Unit SizeCase