Polypropylene Centrifuge Tubes, Sterile
Actual product may vary
Polypropylene Centrifuge Tubes, Sterile
Manufactured from clear polypropylene for superior chemical resistance. Corning™ Polypropylene Centrifuge Tubes are an ideal solution for centrifuge needs.
Have Questions?
Catalog NumberCapacity (Metric)Max. RCFClosure Type
FIS07-200-621500 mL6000 x gPlug
Catalog number FIS07-200-621
Price (SGD)
117.00
Pack
Capacity (Metric):
500 mL
Max. RCF:
6000 x g
Closure Type:
Plug
Price (SGD)
117.00
Pack
Manufactured from clear polypropylene for superior chemical resistance. Corning™ Polypropylene Centrifuge Tubes are an ideal solution for centrifuge needs.
  • Corning centrifuge tubes feature black printed graduations and a large white marking spot.
  • Available with your choice of cap styles: the advanced Corning CentriStarTM cap or the original plug seal cap.
  • Sterile. 
  • Nonpyrogenic.
  • 95 kPA (14 psi) pressure tested – plug seal cap only.
PP = polypropylene, PET = polyethylene terephthalate, RCF = relative centrifugal force (x g).
Specifications
Certifications/ComplianceCertified non-pyrogenic
Closure MaterialHDPE
Closure ColorOrange
ColorNatural
Capacity (Metric)500 mL
MaterialPolypropylene
No. per Case36
No. per Pack6
PackagingNo Rack
SterilitySterile
ShapeConical
Capacity (English)16.9 oz.
Bottom ShapeConical
Closure TypePlug
Writing AreaNo
GraduatedYes
Product TypeCentrifuge Tube
Max. RCF6000 x g
Unit SizePack