Costar™ Sterile Disposable Reagent Reservoirs
Costar™ Sterile Disposable Reagent Reservoirs
Actual product may vary
Costar™ Sterile Disposable Reagent Reservoirs
Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Packaging
FIS07-200-12750 mLBulk
FIS07-200-12850 mLIndividually Wrapped
FIS07-200-129100 mLBulk
Catalog number FIS07-200-127
Price (SGD)
710.00
CS
Add to cart
Capacity (Metric):
50 mL
Packaging:
Bulk
Price (SGD)
710.00
CS
Add to cart
Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.
Costar™ reagent reservoirs are ideal for repetitively filling multichannel pipettors.
  • Manufactured from modified polystyrene (white)
  • Individually wrapped, sterile
  • Disposable
Specifications
Capacity (Metric)50 mL
Dimensions (L x W x H)145 x 70 x 28 mm
MaterialPolystyrene
SterilitySterile
TypeReagent Reservoir
Height (Metric)28 mm
Length (Metric)145 mm
No. per Case200/Cs.
No. per Pack5/Pk.
PackagingBulk
Width (Metric)70 mm