Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Racks
▸
Tube Racks
▸
Tube Supports for Nessler Color Tubes
Actual product may vary
Bennett Wood Specialties
Tube Supports for Nessler Color Tubes
Have Questions?
Catalog Number
Positions
Product Type
For Use With (Equipment)
FIS07-060A
12
Tall Form Tube Rack
50mL Nessler color tubes
Catalog number
FIS07-060A
Price (SGD)
368.00
Each
Add to cart
Positions:
12
Product Type:
Tall Form Tube Rack
For Use With (Equipment):
50mL Nessler color tubes
Price (SGD)
368.00
Each
Add to cart
Specifications
Autoclavable
Not Autoclavable
Holds
Nessler Tubes (50 mL)
Material
Wood
Positions
12
For Use With (Equipment)
50mL Nessler color tubes
Product Type
Tall Form Tube Rack