Kimble™ Exax Brand Nessler Tall-Form Color Comparison Tubes with standard taper Cap Stoppers
Kimble™ Exax Brand Nessler Tall-Form Color Comparison Tubes with standard taper Cap Stoppers
Actual product may vary
DWK Life Sciences
Kimble™ Exax Brand Nessler Tall-Form Color Comparison Tubes with standard taper Cap Stoppers
With standard taper Cap Stoppers
Have Questions?
Change viewbuttonViewtableView
Catalog NumberGraduationsStandard Taper Joint
FIS07-059A50 mL19/10
FIS07-059-1B50 and 100 mL24/12
Catalog number FIS07-059A
Price (SGD)
1,290.00
CS
Add to cart
Graduations:
50 mL
Standard Taper Joint:
19/10
Price (SGD)
1,290.00
CS
Add to cart
With standard taper Cap Stoppers
  • Stopper has shadowless bottom so it need not be removed when making readings
  • The word “shadowless” is printed on side of all caps to distinguish them from regular weighing stoppers
  • 50 to 100mL size designed from ASTM D 1209-00 (Kimble 45325A-50; 45325B-50100)
Specifications
MaterialBorosilicate Glass
Volume (Metric)50 mL
ColorColorless
Closure TypeStandard Taper Stopper
Length (Metric)312 mm
Diameter (Metric) Outer20 mm
Certifications/ComplianceNNN-T-175, USP, ASTM E 438, APHA
For Use With (Application)Used to fill with standard solution colors for visual color comparison with similar tubes containing solution sample
IncludesWhite ceramic enamel rings and legend
Graduations50 mL
Standard Taper Joint19/10
Scale TypeGraduated Ring
Length (English) Scale7.87 to 8.66 in.
Unit SizeCS