▸
Tubes
▸
Centrifuge Tubes
▸
Leucosep Centrifuge Tubes with Porous Barrier
Actual product may vary
Greiner Bio-One
Leucosep Centrifuge Tubes with Porous Barrier
Efficient separation of lymphocytes and mononuclear cells.
Have Questions?
Change view
Catalog Number
Length (Metric)
Bottom Shape
FIS07-000-983
115 mm
Conical
FKS15805547
115 mm
Conical
2 Options
Catalog number
FIS07-000-983
Price (SGD)
448.00
Case
Add to cart
Length (Metric):
115 mm
Bottom Shape:
Conical
Price (SGD)
448.00
Case
Add to cart
Non-cytotoxic, non-pyrogenic
Enrichment directly from whole blood
Simplified filling through porous barrier
Rapid separation in 15 minutes at room temperature
No additional laboratory equipment required, asepticall
Specifications
Closure Color
Natural
Color
Translucent; print of graduation: blue
Capacity (Metric)
50 mL
Material
Polypropylene
Sterility
Sterile
Length (Metric)
115 mm
Bottom Shape
Conical
Closure Included
Yes
Graduated
Yes
Product Type
Centrifuge Tube