▸
Dishes
▸
Petri Dishes
▸
Polystyrene Vented Petri Dishes
Actual product may vary
Greiner Bio-One
Polystyrene Vented Petri Dishes
Used for easy stacking. Greiner Bio-One Polystyrene Vented Petri Dishes is designed for improved gas exchange.
Have Questions?
Change view
Catalog Number
Sterility
Diameter (Metric)
Height (Metric)
FIS07-000-339
Sterile
60 mm
15 mm
FIS07-000-335
Sterile
94 mm
16 mm
2 Options
Catalog number
FIS07-000-339
Price (SGD)
543.00
Case
Add to cart
Sterility:
Sterile
Diameter (Metric):
60 mm
Height (Metric):
15 mm
Price (SGD)
543.00
Case
Add to cart
Vented for improved gas exchange
Manufactured from crystal polystyrene for optimal analysis by microscopy
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
Specifications
Color
Clear
Material
Polystyrene
Shape
Round
Sterility
Sterile
Type
Petri Dish
No. per Case
600
Vent
Yes
Dimensions (Dia. x H)
60 x 15 mm
Height (Metric)
15 mm
Diameter (Metric)
60 mm