Kimtech Science™ Precision Wipes™ Tissue Wipers
Actual product may vary
Fisherbrand
Kimtech Science™ Precision Wipes™ Tissue Wipers
Ideal for general low-lint cleaning tasks
Have Questions?
Catalog NumberDimensions (L x W)Packaging QuantityPly
FIS06-677-708.4 x 4.4 in.286 Wipers/Pk.1-ply
Catalog number FIS06-677-70
Price (SGD)
-
Dimensions (L x W):
8.4 x 4.4 in.
Packaging Quantity:
286 Wipers/Pk.
Ply:
1-ply
  • Pop-Up™ Dispenser Box reduces lint and electrostatic discharge
  • Coated package protects wipes from spills
  • White
Specifications
Product TypeDelicate Task Wipe
Length (English)8.4 in.
Length (Metric)21.33 cm
Width (English)8.4 in.
Width (Metric)11.17 cm
MaterialCellulosic Pulp
ColorWhite
DisposableSingle-use
Dimensions (L x W)8.4 x 4.4 in.
FormatPOP-UP™ box
For Use With (Application)Ideal for general low lint cleaning task
Packaging Quantity286 Wipers/Pk.
Ply1-ply
Unit SizePack