Wheaton™ E-Z Decappers
Wheaton™ E-Z Decappers
Actual product may vary
DWK Life Sciences
Wheaton™ E-Z Decappers
With ergonomically designed handles for sealing and decapping standard laboratory vials
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS06-451-46213 mm Seals
Catalog number FIS06-451-462
Price (SGD)
1,551.00
Case
Add to cart
For Use With (Equipment):
13 mm Seals
Price (SGD)
1,551.00
Case
Add to cart
Wheaton E-Z Decappers are recommended for decapping standard laboratory vials.
  • Several sizes of decapper are available, including those that remove seals 8mm, 11mm, 13mm and 20mm wide.
  • The decappers come with visible size labels for easy identification.
  • User comfort is preserved by an ergonomically designed, cushioned handle. Hand fatigue is reduced by the short handle and redesigned spring assembly.
  • Decappers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Specifications
For Use With (Equipment)13 mm Seals
Unit SizeCase