Check Order Status
Wheaton™ Reusable PVC Coliwasas and Accessories
Actual product may vary
DWK Life Sciences
Wheaton™ Reusable PVC Coliwasas and Accessories
For a variety of applications
Have Questions?
Catalog Number
Product Type
Length (Metric)
Length (English)
FIS06-451-452
Extension and Connector
1.21 m
4 ft.
Catalog number
FIS06-451-452
Price (SGD)
-
Product Type:
Extension and Connector
Length (Metric):
1.21 m
Length (English):
4 ft.
Specifications
Product Type
Extension and Connector
Capacity (English)
25.6 oz.
Capacity (Metric)
760 mL
For Use With (Equipment)
Clear PVC reusable coliwasas
Length (English)
4 ft.
Length (Metric)
1.21 m