Wheaton™ HDPE Serum Bottles
Wheaton™ HDPE Serum Bottles
Actual product may vary
DWK Life Sciences
Wheaton™ HDPE Serum Bottles
Wheaton Serum bottle is manufactured from HDPE and widely used for animal health products and storage of non-medical products.
Have Questions?
Catalog NumberCapacity (Metric)
FIS06-448T120 mL
Catalog number FIS06-448T
Price (SGD)
573.00
CS
Add to cart
Capacity (Metric):
120 mL
Price (SGD)
573.00
CS
Add to cart
  • Manufactured from high density polyethylene
  • Widely used for animal health products and storage of non-medical products
  • Accepts standard rubber stoppers and aluminum seals
Specifications
Capacity4.05 oz., 120 mL
Capacity (Metric)120 mL
Capacity (English)4.05 oz.
ColorNatural
Diameter (English)1.85 in.
Diameter (English) Inner Neck0.78 in.
Diameter (Metric) Outer Neck20 mm
Diameter (Metric) Inner Neck13 mm
MaterialHDPE
Height (Metric)103 mm
Product TypeSerum Bottle
Height (English)4.05 in.
Closure MaterialRubber
Diameter (Metric)47 mm
Unit SizeCS