PYREX™ Reusable Media Storage Bottles
PYREX™ Reusable Media Storage Bottles
Actual product may vary
Fisherbrand
PYREX™ Reusable Media Storage Bottles
Ideal for media, buffers, aqueous and nonaqueous solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)MouthQuantity
FIS06-423B50 mLNarrow10/Cs.
FIS06-423A25 mLNarrow10/Cs.
FIS06-414-1A100 mLNarrow10/Cs.
FIS06-414-1B250 mLWide10/Cs.
FIS06-414-1C500 mLWide10/Cs.
FIS06-414-1D1 LWide10/Cs.
FIS06-414-1E2000 mLWide1/Cs.
FIS06-414-1F5000 mLWide1/Cs.
Catalog number FIS06-423B
Price (SGD)
379.00
CS
Add to cart
Capacity (Metric):
50 mL
Mouth:
Narrow
Quantity:
10/Cs.
Price (SGD)
379.00
CS
Add to cart
Ideal for media, buffers, aqueous and nonaqueous solutions
Heavy duty bottle which can be used for storage as well as mixing and sampling. Includes linerless, one-piece autoclavable polypropylene plug seal cap with drip free pouring rings. Optional red PBT caps and red ETFE pouring rings are available for dry heat sterilization (180°C). Glass bead indicates full capacity line on 100 mL through 2L size. Bottles have permanent white enamel graduations and marking spots. Color-coded polypropylene caps are available in a choice of orange, green, purple, red, and light gray.
Caution: Bottles should NOT be used with bottle top filter units, or in other applications involving vacuum pressure, as breakage may occur. DO NOT have caps tightened immediately after autoclaving, as the vacuum resulting from cooling can cause breakage. A pouring ring is not included on 25 mL size. A vented cap is NOT recommended for use on the 5L and 10L 1395 series bottle.
Specifications
Capacity1.69 oz., 50 mL
Capacity (Metric)50 mL
Capacity (English)1.69 oz.
Diameter (English)1.8 in.
MaterialBorosilicate Glass
No. per Case10/Cs.
No. per Pack10/Pk.
Quantity10/Cs.
SterilityNon-sterile
Closure ColorOrange
Graduations20 to 50 mL
MouthNarrow
ShapeCylindrical
Height (Metric)88 mm
Height (English)3.4 in.
Closure MaterialPolypropylene
Closure SizeGL 32
Diameter (Metric)46 mm
Closure TypePlug Seal
Unit SizeCS