Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Actual product may vary
DWK Life Sciences
Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
Have Questions?
Change view
Catalog Number
Color
Closure Size
FIS06-406-14B
Natural
20 mm
FIS06-406-14A
Natural
13 mm
2 Options
Catalog number FIS06-406-14B
Price (SGD)
341.00
CS
Add to cart
Color:
Natural
Closure Size:
20 mm
Price (SGD)
341.00
CS
Add to cart
Good barrier properties
Color selection includes natural, red, blue and green
Chemically resistant to alcohol and acetone, among others