1x1 image pixel for data collection
Wheaton™ Graduated Media/Lab Bottles: Clear Glass, With Rubber-Lined Cap (Autoclavable)
Wheaton™ Graduated Media/Lab Bottles: Clear Glass, With Rubber-Lined Cap (Autoclavable)
Actual product may vary
DWK Life Sciences
Wheaton™ Graduated Media/Lab Bottles: Clear Glass, With Rubber-Lined Cap (Autoclavable)
Rugged space-savers
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FIS06-404B250 mL33-430 mm
Catalog number FIS06-404B
Price (SGD)
30.00
EA
Estimated availability date 22-Jul-2025
Add to cart
Capacity (Metric):
250 mL
Closure Size:
33-430 mm
Price (SGD)
30.00
EA
Add to cart
Rugged space-savers
  • Clear borosilicate glass bottle with graduations and writing patch
  • Heavy walls prevent breakage
  • Sturdier than Erlenmeyer flasks
  • Smaller diameter at base conserves space in sterilizers and freezers
  • Large opening, special lip, sloping shoulders
  • Graduated in mL to show approximate volumes
  • Hand grips on 1L bottle provide easy handling
Specifications
Capacity8.45 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8.45 oz.
ColorClear
Diameter (English)2.63 in.
MaterialGlass
Closure ColorBlack
Certifications/ComplianceUSP Type 1
GraduatedYes
Height (Metric)152 mm
Height (English)5.98 in.
Closure LinerRubber
Closure MaterialPhenolic
Closure Size33-430 mm
Diameter (Metric)67 mm
Closure TypeScrew Cap
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link