1x1 image pixel for data collection
Castaloy™ Three-Prong Extension Clamps
Castaloy™ Three-Prong Extension Clamps
Actual product may vary
Castaloy™ Three-Prong Extension Clamps
Hold irregular shapes at any angle, surely and safely.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize (Metric) Grip
FIS057698Q8.9 cm
FIS05-769-6Q2.5 cm
FIS05-769-7Q5.71 cm
FIS05-769-8Q8.9 cm
Catalog number FIS057698Q
Price (SGD)
-
Size (Metric) Grip:
8.9 cm
Sturdy extension clamps allow you to hold objects with irregular shapes at any angle, safely and securely.
  • Two separate adjustment screws ensure even, precise pressure on glass and minimize chance of breakage
  • Clamps are easily adjustable to vary holding angle, distance from stand
Specifications
Quantity1 Ea.
TypeExtension Clamp
IncludesFiberglass and Vinyl sleeve
Length (English)10.5 in.
Length (Metric)27 cm
SizeExtension arm O.D.: 0.50 in. (1.3 cm)
Size (Metric) Grip8.9 cm
Size (English) Grip3.5 in.
Jaw Type3-Prong
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link