1x1 image pixel for data collection
Wide-Mouth HDPE Packers with Closure
Wide-Mouth HDPE Packers with Closure
Actual product may vary
Wide-Mouth HDPE Packers with Closure
Wide-Mouth HDPE Packers with Closure
Wide-Mouth HDPE Packers with Closure
Thermo Scientific
Wide-Mouth HDPE Packers with Closure
Choose these containers for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)LevelNo. per Case
FIS05719149D1000 mLCertified12
FIS05-721-132120 mLUnprocessed48
FIS05-721-133250 mLUnprocessed24
FIS05-721-138250 mLUnprocessed280
FIS05-721-139500 mLUnprocessed150
FIS05-721-1351000 mLUnprocessed12
FIS05-721-1401000 mLUnprocessed90
FIS05-721-1242000 mLCertified6
Catalog number FIS05719149D
Price (SGD)
235.00
CS
Add to cart
Capacity (Metric):
1000 mL
Level:
Certified
No. per Case:
12
Price (SGD)
235.00
CS
Add to cart
Choose these containers for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes. Thermo Scientific™ Wide-Mouth HDPE Packers enable easy filling and sample retrieval. Available in small quantity lab packs or higher economy bulk packs. Certified to meet EPA Performance Based Specifications for metals, cyanide and fluoride analyses or unprocessed and ready for your own cleaning procedure.
  • HDPE container with white LDPE-lined Polypropylene Cap
  • Caps assembled onto containers
  • Wide mouth for easy filling and sample retrieval
  • Recommended for collecting liquid, semi-liquid and viscous samples
  • Available bulk packed for added economy
Certified containers come with the following features:
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each container individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
Unprocessed container comes assembled and ready for your own cleaning procedure
Specifications
Capacity32 oz., 1000 mL
Capacity (Metric)1000 mL
Capacity (English)32 oz.
Certification ParametersMetals and water quality, Cyanide, Fluoride
ColorNatural
DescriptionHDPE Packer
Diameter (English) Inner1.88 in.
Diameter (English)3.66 in.
Diameter (Metric) Inner4.78 cm
MaterialHDPE
No. per Case12
PackagingCase of 12
Replacement Closure067-6300WBP
SterilityNon-sterile
TransparencyTranslucent
CertifiedMetals; Cyanide; Fluoride; Other inorganics
Closure ColorWhite
LevelCertified
Volume (English) Brim35.5 oz.
Volume (Metric) Brim1051 mL
Certifications/ComplianceCertified containers meet or exceed EPA performance-based standards for metals, cyanide, and fluoride; manufactured under an ISO certified quality management system.
ShapeRound
Height (Metric)178 mm
Height (English)7 in.
Closure LinerFoam/LDPE
Closure MaterialPolypropylene
Closure Size63-400 mm
Diameter (Metric)93 mm
Unit SizeCS
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link