Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Education Supplies
▸
General Classroom Products
▸
Writing Implements and Accessories
▸
Diamond-Tip Pencil
Krackeler Scientific
Diamond-Tip Pencil
Have Questions?
Catalog Number
Tip Material
FIS056001572N
Diamond
Catalog number
FIS056001572N
Price (SGD)
-
Tip Material:
Diamond
Specifications
Product Type
Pencil
Tip Material
Diamond
For Use With (Application)
Used for clean and easy cuts in fused Silica, Glass and Aluminum-clad capillary columns