Hoffman Open-Side Tubing Clamps
Actual product may vary
Humboldt
Hoffman Open-Side Tubing Clamps
For flow regulation in flexible tubing
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeHeight (English)Width (English)
FIS05-875BLarge0 to 0.62 in.1 in.
FIS05-875ASmall0 to 0.5 in.0.75 in.
Catalog number FIS05-875B
Price (SGD)
16.00
Each
Size:
Large
Height (English):
0 to 0.62 in.
Width (English):
1 in.
Price (SGD)
16.00
Each
  • One side is open for easy insertion of tubing without disassembly
  • Nickel-plated brass
  • Resist corrosion
  • Won't rust
Specifications
QuantityEach
TypeTubing Clamp
MaterialNickel-plated Brass
For Use With (Application)Control the flow in flexible tubing
Height (English)0 to 0.62 in.
SizeLarge
Width (English)1 in.
Jaw TypeOpen
Unit SizeEach