Hoffman Tubing Clamps
Hoffman Tubing Clamps
Actual product may vary
Humboldt
Hoffman Tubing Clamps
Regulate flow from fully open to closed
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSizeHeight (English)Height (Metric)Jaw Type
FIS05-871A0.75 in.0 to 0.62 in.0 to 16 mmSwivel
FIS05-871B1 in.0 to 0.75 in.0 to 19 mmSwivel
FIS05-870B1 in.0 to 0.75 in.0 to 19 mmFixed
Catalog number FIS05-871A
Price (SGD)
14.00
Each
Add to cart
Size:
0.75 in.
Height (English):
0 to 0.62 in.
Height (Metric):
0 to 16 mm
Jaw Type:
Swivel
Price (SGD)
14.00
Each
Add to cart
Regulate flow from fully open to closed
  • For flexible tubing
  • Nickel-plated brass
  • Won't rust
  • Lower jaw is either fixed or swivel-mounted for easy insertion of tubing without disconnecting apparatus
Specifications
Quantity1 Ea.
TypeTubing Clamp
MaterialNickel-plated Brass
For Use With (Equipment)Control The Flow in Flexible Tubing
For Use With (Application)Control the flow in flexible tubing
Height (Metric)0 to 16 mm
Height (English)0 to 0.62 in.
Size0.75 in.
Jaw TypeSwivel
Unit SizeEach