Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Clamps and Supports
▸
Clamps
▸
Flatjaw Pinchcocks
Actual product may vary
Humboldt
Flatjaw Pinchcocks
One-piece rust resitant brass clamp
Have Questions?
Catalog Number
Size
Length (English)
Length (Metric)
FIS05-850B
Large
3.25 in.
8.25 cm
Catalog number
FIS05-850B
Price (SGD)
9.00
Each
Add to cart
Size:
Large
Length (English):
3.25 in.
Length (Metric):
8.25 cm
Price (SGD)
9.00
Each
Add to cart
One-piece rust resitant brass clamp
Nickel-plated
Clamp flexible tubing up to 0.62 in. (16mm)
Specifications
Type
Mohr Pinchcock
Material
Nickel, Steel
Length (English)
3.25 in.
Length (Metric)
8.25 cm
Size
Large