▸
Clamps and Supports
▸
Clamps
▸
Swing Jaw™ Tubing Clamps
Actual product may vary
Bel-Art
Swing Jaw™ Tubing Clamps
Provide nonslip closures for flexible tubing
Have Questions?
Catalog Number
Diameter (Metric) Outer Tubing
FIS05-834-5
20 mm
Catalog number
FIS05-834-5
Price (SGD)
-
Diameter (Metric) Outer Tubing:
20 mm
Made of zinc-plated steel
Knurled adjusting nut bears on a swinging jaw which permits full opening without removal
Removable from tubing without disconnecting apparatus
Smooth bearing surfaces will not damage tubing; tubing cannot slip out of clamp
Strong enough to use with heavy wall vacuum lines
Specifications
Type
Tubing Clamp
Material
Zinc-plated Steel
For Use With (Equipment)
20 mm (3/4 in.) O.D. flexible tubing
Diameter (English) Outer Tubing
3/4 in.
Diameter (Metric) Outer Tubing
20 mm
Color
Silver
Jaw Type
Swing