BioMax™ Light Film
Actual product may vary
Carestream Health
BioMax™ Light Film
Developed specifically for the detection of chemiluminescent probes
Have Questions?
Catalog NumberDimensions (LxW)Product TypeQuantity
FIS05-728-558 x 10 in.BioMax Light-1 Film: Noninterleaved50 Sheets/Pk., Bulk Pack
Catalog number FIS05-728-55
Price (SGD)
-
Dimensions (LxW):
8 x 10 in.
Product Type:
BioMax Light-1 Film: Noninterleaved
Quantity:
50 Sheets/Pk., Bulk Pack
  • Produces high signal-to-noise ratio
  • Coated on both sides of a clear base
  • Can be used for direct autoradiography and intensifying screen exposures with commonly used isotopes, but requires longer exposure times than BioMax MS, and BioMax XAR™ film
  • Suitable for manual and automated processing
Kodak autoradiography and electron micrography films should be handled under proper safelight conditions using the Kodak Dark Room Illuminator (Mfr. No. 1132752 ). This self-contained unit is matched for use with all autoradiography and electron micrography films.Use of other safelights with improper filters or improper wattage bulbs will result in increased background fog on the exposed film and poorer images.
Specifications
Product TypeBioMax Light-1 Film: Noninterleaved
For Use With (Application)For manual and automated processing
Quantity50 Sheets/Pk., Bulk Pack
Dimensions (LxW)8 x 10 in.
Unit SizePack