Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle with Closure
Actual product may vary
Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle with Closure
Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle with Closure
Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle with Closure
Thermo Scientific
Nalgene™ Wide-Mouth HDPE Sterile Sample Bottle with Closure
Choose this container, which is break-resistant, guaranteed leakproof and certified sterile, for water sample collection for microbiological analyses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FIS05-719-359B250 mL43 mm
FIS05-719-359A125 mL38 mm
FIS05-719-359C500 mL53 mm
Catalog number FIS05-719-359B
Price (SGD)
1,046.00
Case
Capacity (Metric):
250 mL
Closure Size:
43 mm
Price (SGD)
1,046.00
Case
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Sterile Sample Bottles are recommended for collecting water samples for microbiological analyses. The icon of quality, Nalgene containers are guaranteed to be leakproof† and break resistant. Durable construction withstands the rigors of field operations. Wide mouth for easy aseptic filling. Certified sterile.
  • HDPE container with leakproof† HDPE closure
  • Leakproof† guaranteed
  • Certified Sterile
  • Caps assembled onto containers and individually shrink-wrapped for tamper evidence and sterility assurance
  • Packaged in shrink-wrapped trays
  • Wide mouth for easy aseptic filling
  • Recommended for collecting water samples for microbiological analyses
Certified containers come with the following features:
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each container individually barcoded with lot number and unique container number
  • Each closure individually sealed with tamper-evident shrink wrap
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
Note:
†The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity8 oz., 250 mL
Capacity (Metric)250 mL
Capacity (English)8 oz.
Certification ParametersSterile, Metals and water quality, Cyanide, Fluoride
ColorNatural
DescriptionNalgene WM HDPE Bottle
Diameter (English) Inner1.31 in.
Diameter (English)2.4 in.
Diameter (Metric) Inner3.33 cm
IncludesCertified containers come with the following features: Certified sterile Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability; Each container individually barcoded with lot number and unique container number; E
MaterialHDPE
No. per Case60
PackagingCase of 60
Replacement ClosureNA
SterilitySterile
TransparencyTranslucent
Closure ColorWhite
LevelCertified
Volume (English) Brim10.1 oz.
Volume (Metric) Brim300 mL
Certifications/ComplianceCertified Sterile; Certified containers meet or exceed EPA performance-based standards for metals, cyanide, and fluoride; manufactured under an ISO certified quality management system.
Height (Metric)131 mm
Height (English)5.15 in.
Closure MaterialHDPE
Closure Size43 mm
Diameter (Metric)61 mm
Unit SizeCase