I-Chem™ Wide-Mouth Amber Glass Packer with Closure
Actual product may vary
I-Chem™ Wide-Mouth Amber Glass Packer with Closure
I-Chem™ Wide-Mouth Amber Glass Packer with Closure
I-Chem™ Wide-Mouth Amber Glass Packer with Closure
Thermo Scientific
I-Chem™ Wide-Mouth Amber Glass Packer with Closure
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required with these wide-mouth, amber glass packers.
Have Questions?
Catalog NumberCapacity (Metric)LevelClosure SizeNo. per Case
FIS05-719-28860 mLProcessed33-400 mm24
Catalog number FIS05-719-288
Price (SGD)
365.00
Case
Capacity (Metric):
60 mL
Level:
Processed
Closure Size:
33-400 mm
No. per Case:
24
Price (SGD)
365.00
Case
Collect soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required with Thermo Scientific™ I-Chem™ Wide-Mouth Amber Glass Packers. Packers are available Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals anlyses, Processed the same way as the certified jars but without certification documentation, or Unprocessed and ready for your own cleaning procedure.
  • Amber Type III soda-lime glass jar; recommended for light-sensitive samples
  • PTFE-lined white polypropylene closed-top cap
  • Sizes range from 60 to 2500mL
  • Closures assembled on bottles
Certified jars come with the following features:
  • Certified to meet or exceed EPA performance-based standards for semi-volatile organics, pesticides, PCBs and metals analyses
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each jar individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
  • Chipboard divider packaging to cushion and help prevent breakage; overpack required for shipping
Processed jars are processed in the same manner as certified jars and come with all the features of certified jars, but jars are not barcoded, and CofA is not included.
Unprocessed jars are ready for your own in-house cleaning and certification process. Boxed with chipboard dividers to help prevent breakage; overpack required for shipping.
Specifications
Capacity2 oz., 60 mL
Capacity (Metric)60 mL
Capacity (English)2 oz.
Certification ParametersSemi-VOA, pesticides, PCBs, metals
ColorAmber
DescriptionAmber Glass Packer
Diameter (English) Inner1 in.
Diameter (English)1.73 in.
Diameter (Metric) Inner2.5 cm
MaterialGlass (Type 3)
No. per Case24
Replacement ClosureB167-3300BL
Closure ColorWhite
LevelProcessed
Liner MaterialPTFE
Volume (English) Brim2.4 oz.
Volume (Metric) Brim70 mL
Certifications/ComplianceCertified and Processed jars meet or exceed EPA performance based standards for semi-volatile organics, pesticides, PCB's and metals analysis; manufactured under an ISO certified quality management system.
MouthWide
Height (Metric)75 mm
Product TypeAmber Packer Bottle
Height (English)2.95 in.
Closure MaterialPolypropylene
Closure Size33-400 mm
Diameter (Metric)44 mm
Closure TypeClosed-top Cap
Unit SizeCase