High Impact Polystyrene (HIPS) Drying Rack
High Impact Polystyrene (HIPS) Drying Rack
Actual product may vary
High Impact Polystyrene (HIPS) Drying Rack
High Impact Polystyrene (HIPS) Drying Rack
High Impact Polystyrene (HIPS) Drying Rack
Fits up to 72 pegs.
Have Questions?
Catalog NumberPositions
FIS05-718-4072
Catalog number FIS05-718-40
Price (SGD)
1,147.00
EA
Add to cart
Positions:
72
Price (SGD)
1,147.00
EA
Add to cart
Fits up to 72 pegs.
  • Injection-molded, high-impact, light blue polystyrene
  • Pegs hold cylinders, bottles, beakers and containers etc. and snap in or out easily when desired
  • Comes with 72 large 4 in. long (10.2cm) pegs, expansion hanging hooks, draining tubes and fastening stoppers for coupling with additional racks
  • Interchangeable pegs for use with test tubes available separately
  • Bag of 11 Pegs made of high impact polystyrene
Specifications
Product TypeDrying Rack
AutoclavableNot Autoclavable
ColorWhite
Holds72
MaterialPolystyrene
Positions72
Length (Metric)44.92 cm
Length (English)17.68 in.
Width (Metric)11.11 cm
Width (English)4.375 in.
Height (Metric)63 cm
Height (English)24.82 in.
Unit SizeEA