Tubing Sleeves
Actual product may vary
Tubing Sleeves
Tubing Sleeves
Idex
Tubing Sleeves
Ensure good port connection with capillary tubing
Have Questions?
Catalog NumberFor Use WithColorDiameter (Metric) Inner
FIS05-700-353340-380 μm O.D. TubingGreen395 μm
Catalog number FIS05-700-353
Price (SGD)
-
For Use With:
340-380 μm O.D. Tubing
Color:
Green
Diameter (Metric) Inner:
395 μm
Ensure good port connection with capillary tubing
  • Maintain concentric connection with minimal dead volume
  • Color coded for easy identification
Sleeves are not permanently attached to a ferrule so they can easily adapt to varying tubing pocket depths minimizing dead volume. Sleeves are designed to be used with 1/16 in. OD, 10-32 threaded fittings to adapt capillary tubing to standard coned ports. Sleeves require a wrench tightened nut to achieve proper sealing and withstand temperatures up to 257°F (125°C) and pressures up to 6000 psi (414 bar).
Specifications
TypeSleeve
ColorGreen
Diameter (English) Inner0.015 in.
Diameter (Metric) Inner395 μm
For Use With340-380 μm O.D. Tubing
MaterialPEEK
Unit SizePack