Quick Connect Adapter
Actual product may vary
Quick Connect Adapter
Quick Connect Adapter
Idex
Quick Connect Adapter
Convenience of quick connect and disconnect of a luer connection
Have Questions?
Catalog NumberTypeMaterial
FIS05-700-335Female Luer AdapterPEEK
Catalog number FIS05-700-335
Price (SGD)
-
Type:
Female Luer Adapter
Material:
PEEK
Convenience of quick connect and disconnect of a luer connection
Quick connect threaded luer adapters are designed to connect any male luer to any female luer in a variety of applications. Male luer fittings are available with or without lock hubs. Lock hubs provide added security of holding the connection in place.
Specifications
TypeFemale Luer Adapter
Diameter (English) Inner0.05 in.
MaterialPEEK
ColorTan
For Use With (Equipment)Female Luer x Female 10-32 Coned
Unit SizeEach