SUV39H1 Mouse anti-Human, Mouse, Clone: MG44, MilliporeSigma™
SUV39H1 Mouse anti-Human, Mouse, Clone: MG44, MilliporeSigma™
Actual product may vary
DWK Life Sciences
SUV39H1 Mouse anti-Human, Mouse, Clone: MG44, MilliporeSigma™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FIS05-615
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Anti-SUV39H1 Antibody, clone MG44 is a Mouse Monoclonal Antibody for detection of SUV39H1 also known as KMT1A, Histone H3-K9 Methyltransferase 1 and has been validated in Chimmunoprecipitation, immunoprecipitation and western blotting.
For Research Use Only
Specifications
AntigenSUV39H1
ApplicationsChIP Assay, Immunoprecipitation, Western Blot
ClassificationMonoclonal
CloneMG44
ConjugateUnconjugated
FormulationProtein G Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative.
Gene Accession No.NM_003173.2
Gene SymbolsSUV39H1, Su, SUV39H, MG44
Host SpeciesMouse
ImmunogenMBP fusion protein corresponding to residues 1-412 of human SUV39H1
Purification MethodUnpurified
Quantity200 μL
Primary or SecondaryPrimary
Target SpeciesHuman, Mouse
FormAscites
IsotypeIgG
Unit SizePK
Contents & Storage
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.