Luciferase Mouse anti-All, Clone: clone21, MilliporeSigma™
Luciferase Mouse anti-All, Clone: clone21, MilliporeSigma™
Actual product may vary
DWK Life Sciences
Luciferase Mouse anti-All, Clone: clone21, MilliporeSigma™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FIS05-603
Price (SGD)
1,098.00
CS
Add to cart
Price (SGD)
1,098.00
CS
Add to cart
Detect Luciferase using this Anti-Luciferase Antibody, clone21 validated for use in western blot, immunohistochemistry.
For Research Use Only
Specifications
AntigenLuciferase
ApplicationsImmunohistochemistry, Western Blot
ClassificationMonoclonal
Clone21
ConjugateUnconjugated
Formulation0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Host SpeciesMouse
ImmunogenFull-length recombinant luciferase from Photinus pyralis.
Purification MethodProtein G chromatography
Quantity200 μg
Primary or SecondaryPrimary
Target SpeciesAll species
FormPurified
IsotypeIgG1
Unit SizeCS
Contents & Storage
−20°C for 24 months.