Search Fisher Scientific
| Catalog Number | Nominal Capacity | Max. Force |
|---|---|---|
| FIS05-579-30 | 175 mL | 5800 x g |
Easily visualize samples through the transparent polystyrene of the Thermo Scientific™ Nalgene™ Sterile Conical-Bottom Centrifuge Bottle, an economical and disposable solution for low-speed, large-volume cell harvesting, pelleting and protein purification. Well suited for sterile cell cuture applications these conical-bottom centrifuge bottles are the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors. Bottles are individually wrapped and ready to use.
Requires:
Must be used with Nalgene adapter for conical-bottom centrifuge bottles (DS3126-0175).