▸
Racks
▸
Microtube Racks
▸
Microcentrifuge Tube Rack Cover
Actual product may vary
Fisherbrand
Microcentrifuge Tube Rack Cover
Accommodates extra height of screw-cap microcentrifuge tubes.
Have Questions?
Catalog Number
Positions
FIS05-541-28
96
Catalog number
FIS05-541-28
Price (SGD)
-
Positions:
96
Accommodates extra height of screw-cap microcentrifuge tubes.
Clear cover slides on to stay in place during transportation or storage
Recessed area allows stacking
Specifications
Autoclavable
Not Autoclavable
Color
Clear
Holds
Microcentrifuge Tubes (1.5 to 2 mL)
Material
Polypropylene
Positions
96
Shape
Rectangular
Stackable
No
Sterility
Non-sterile
Quantity
5/Pk.
Length (Metric)
203 mm
Product Type
Microcentrifuge Tube Rack
Shelves
None
Height (Metric)
29 mm
Width (Metric)
119 mm