Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Type | Packaging | Sterility |
|---|---|---|---|---|
| FIS05-539-7 | 50 mL | Plug | Bulk | Sterile |
| FIS05-539-4 | 15 mL | Plug | Sleeve | Sterile |
| FIS07-200-886 | 15 mL | Flat Cap | Sleeve | Sterile |
| FIS05-539-12 | 15 mL | Flat Cap | Racked | Sterile |
| FKS11849650 | 15 mL | Flat Cap | Racked | Sterile |
| FKS11889640 | 15 mL | Plug | Racked | Sterile |
| FIS05-539-13 | 50 mL | Flat Cap | Bulk | Sterile |
| FKS11512303 | 50 mL | Flat Cap | Bulk | Sterile |
| FIS05-539-6 | 50 mL | Plug | Racked | Sterile |
| FKS11809650 | 50 mL | Plug | Bulk | Sterile |
| FIS05-539-8 | 50 mL | Flat Cap | Racked | Sterile |
| FKS11819650 | 50 mL | Flat Cap | Racked | Sterile |