Polypropylene Cushion
Actual product may vary
Polypropylene Cushion
For use with 175 and 225mL Conical centrifuge tube
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS05-538-63175 and 225 mL Conical Centrifuge Tube
Catalog number FIS05-538-63
Price (SGD)
1,329.00
Case
For Use With (Equipment):
175 and 225 mL Conical Centrifuge Tube
Price (SGD)
1,329.00
Case
Cushions and extractor for use with 175 mL and 225 mL conical centrifuge tubes (Cat. Nos. 352075 and 352076). Nonsterile. 8/case.
Material
Cushions: Polypropylene
Wire Extractor: stainless steel
Specifications
TypeCushion
MaterialPolypropylene
For Use With (Equipment)175 and 225 mL Conical Centrifuge Tube
IncludesStainless Steel wire extractor
Unit SizeCase