1x1 image pixel for data collection
TCF-1 Mouse anti-Human, Clone: 7H3, MilliporeSigma™
TCF-1 Mouse anti-Human, Clone: 7H3, MilliporeSigma™
Actual product may vary
TCF-1 Mouse anti-Human, Clone: 7H3, MilliporeSigma™
Mouse Monoclonal Antibody
Have Questions?
Catalog number FIS05-510
Price (SGD)
2,707.00
CS
Add to cart
Price (SGD)
2,707.00
CS
Add to cart
Detect TCF-1 with Anti-TCF-1 Antibody, clone 7H3 (Mouse Monoclonal Antibody), that has been shown to work in immunocytochemistry, immunohistochemistry and western blotting.
For Research Use Only
Specifications
AntigenTCF-1
ApplicationsImmunocytochemistry, Immunohistochemistry, Western Blot
ClassificationMonoclonal
Clone7H3
ConjugateUnconjugated
FormulationProtein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.5, 0.15M NaCl and 0.05% sodium azide
Gene Accession No.NM_003202
Gene SymbolsTCF1, MODY3, TCF-1, HNF1, HNF-1A, HNF1A, LFB1, HNF1a
Host SpeciesMouse
ImmunogenMaltose binding protein (MBP) fusion protein corresponding to residues 102-269 of human TCF-1A splice variant. The epitope maps to a region just N-terminal to the High Mobility Group (HMG) box
Purification MethodProtein G purfied
Quantity100 μg
Primary or SecondaryPrimary
Target SpeciesHuman
FormPurified
IsotypeIgG
Unit SizeCS
Contents & Storage
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link