Orbital and Reciprocating Shaking Water Bath Accessories: Gable Cover
Actual product may vary
Boekel Scientific
Orbital and Reciprocating Shaking Water Bath Accessories: Gable Cover
For use with Boekel Scientific Orbital and Reciprocating Shaking Water Baths.
Have Questions?
Catalog NumberDepth (English)
FIS05-450-11112.6 in.
Catalog number FIS05-450-111
Price (SGD)
1,182.00
Each
Depth (English):
12.6 in.
Price (SGD)
1,182.00
Each
  • Stainless steel
Specifications
For Use With (Equipment)24L Shaking water bath
TypeGable Cover
MaterialStainless Steel
Width (English)20.4 in.
Width (Metric)52 cm
Height (English)3.6 in.
Height (Metric)9.2 cm
Depth (English)12.6 in.
Depth (Metric)32 cm
Shipping Weight (Metric)5.8 kg
Weight (English)5.5 lb.
Weight (Metric)2.5 kg
Unit SizeEach