Conical Tubes with SnapTec™ Cap
Actual product may vary
Conical Tubes with SnapTec™ Cap
Conical Tubes with SnapTec™ Cap
Conical Tubes with SnapTec™ Cap
Conical Tubes with SnapTec™ Cap
Fisherbrand
Conical Tubes with SnapTec™ Cap
Eppendorf™ Conical Tubes with SnapTec™ Cap offer secure sample containment for diverse applications. The innovative SnapTec™ Cap ensures easy, secure sealing, making them ideal for molecular biology, genomics, and more. The conical design aids optimal sample recovery, enhancing efficiency and reliability in your lab.
Have Questions?
Catalog NumberCapacity (Metric)Purity or Quality GradeColorPackaging
FIS05-414-07345 mLPyrogen-, DNase-, RNase-, Human and Bacterial DNA-freeClear10 Bags x 20 Tubes
Catalog number FIS05-414-073
Price (SGD)
453.00
Each
Capacity (Metric):
45 mL
Purity or Quality Grade:
Pyrogen-, DNase-, RNase-, Human and Bacterial DNA-free
Color:
Clear
Packaging:
10 Bags x 20 Tubes
Price (SGD)
453.00
Each
  • SnapTec™ Cap affords effortless, secure sealing for sample integrity in molecular biology and genomics.
  • Conical design enhances recovery, crucial for applications like PCR, DNA/RNA isolation, and centrifugation.
  • Well-suited for various lab tasks, ensuring adaptability across experiments.
  • Durable construction crafted for longevity, supporting applications like cell culture, protein purification, and more.
  • Streamlines processes, benefitting applications such as qPCR, cloning, and sample storage.
  • Available in 25 mL and SnapTec™ 50 to 45 mL formats.
  • Free of plasticizers, biocides or mold release agents
Specifications
Closure MaterialPolypropylene
ColorClear
Capacity (Metric)45 mL
MaterialPolypropylene
No. per Pack20 Pk.
Packaging10 Bags x 20 Tubes
Quantity200 Pk.
SterilitySterile
AutoclavableAutoclavable
ShapeCylindrical
Clean ClaimsSterile, pyrogen-, DNase-, RNase-, human and bacterial DNA-free
Closure TypeSnapTec Cap
Writing AreaYes
Temperature Range (Metric)-86°C to -100°C
Diameter (Metric)30 mm
Product TypeConical Tube
Max. RCF17,000 x g
Purity or Quality GradePyrogen-, DNase-, RNase-, Human and Bacterial DNA-free
Unit SizeEach