▸
Tubes
▸
Microtubes
▸
Premium Microcentrifuge Tubes: 1.5mL
Actual product may vary
Premium Microcentrifuge Tubes: 1.5mL
Available in eight colors or in assorted-color packs.
Have Questions?
Change view
Catalog Number
Color
FIS05-408-129
Natural
FIS05-408-130
Red
FIS05-408-132
Blue
FIS05-408-133
Green
FIS05-408-131
Yellow
FIS05-408-134
Amber
FIS05-408-135
Orange
FIS05-408-136
Lavender
FIS05-408-137
Assorted
9 Options
Catalog number
FIS05-408-129
Price (SGD)
52.00
Pack
Add to cart
Color:
Natural
Price (SGD)
52.00
Pack
Add to cart
Flat-top snap caps provide a safe, liquid-tight, reliable seal even with prolonged boiling
Capacity: 1.5mL
High-clarity polypropylene
Graduated in 0.1 and 0.25mL markings
Withstand forces up to 30,000×G
Easy-open caps with needle insertion spot and textured marking area
Labeling area on tube exterior
Highly polished interior for maximum sample recovery
Temperature range of -80° to +121°C
Fit all standard rotors
Nonpyrogenic
Autoclavable at 121°C/15psi for 15 minutes
Specifications
Cap Style
Snap Cap with Flat Top
Certifications/Compliance
RNase, DNase-free
Autoclavable
Autoclavable
Capacity (Metric)
1.5 mL
Color
Natural
Material
Polypropylene
Includes
Needle insertion spot
Label Type
Labeling Area
Sterility
Non-sterile
Graduated
Yes
Closure Type
Flat Snap Cap
Max. RCF
30,000 x g
Diameter (Metric)
10.8 mm
Length (Metric)
40.6 mm